VWR® Burette, Rotaflow, PTFE Stopcock, Class A
Supplier: VWR Collection
VWRABURPA-10EA 11000 INR
VWRABURPA-10 VWRABURPA-25 VWRABURPA-50 VWRABURPA-100
VWR® Burette, Rotaflow, PTFE Stopcock, Class A
Burettes
Burettes fitted with rotaflow general purpose screw thread stopcock with PTFE key.
Certifications: IS 1997:2008, ISO 385:2005.