Liquid Nitrogen Container, High Capacity
Supplier: Haier
HAIEYDS-10-125-FEA 60999 INR
HAIEYDS-10-125-F
Liquid Nitrogen Container, High Capacity
Dewars and Cryogenic Vessels
High capacity series for storage or transport (round canisters) provides two cryopreservation solutions for long-term static storage and transportation of biological samples.
  • High strength, light weight and small size aluminum construction
  • Ultra-low evaporation loss
  • Numbered index for canister position
  • High thermal efficiency
  • Heavy duty lockable enclosure offers excellent security
  • 5 year vacuum warranty