Chloramphenicol 97-103% USP

Supplier: VWR Chemicals
Danger

Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

0230-100GEA 0 INR
0230-100G 0230-500G
Chloramphenicol 97-103% USP
Chloramphenicol
Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1,49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now

Specification Test Results

Chromatographic Purity Test Pass
Melting Range 149 °C - 153 °C
pH (2.5%, Water) @25°C 4.5 - 7.5
Purity 97 % - 103 %
Specific Rotation 17 °- 20 °


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR