Silver sulphate ≥99% AR ACS, Rankem®

Supplier: RANKEM
Danger

Synonyms: Disilver sulphate

S0664 S0663
RANKS0664EA 62617.5 INR
RANKS0664 RANKS0663
Silver sulphate ≥99% AR ACS, Rankem®
Silver sulphate

High purity reagents that meet or exceed the ACS reagent Specifications.


  • Batch-to-batch consistency
  • Tested for heavy metals
  • Suitable for advanced chemical testing
  • Available in various pack size


White crystalline powder.

Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 3077
ADR: 9 III
Merck Index: 13,08603

Order Now

Specification Test Results

Guarantee Analysis .
Assay (Ag2SO4) Min. 99.00 %
Identity Passes test
Maximum Limits Of Impurities .
Insoluble Matter And Silver Chloride Max. 0.02 %
Substance Not Ppt. By HCl Max. 0.03 %
Iron (Fe) ≤ 0.001%
Nitrate (NO3) ≤ 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR