Silver nitrate ≥99.8% LR, Rankem®

Supplier: RANKEM
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

S0090 S0080
RANKS0090EA 41382 INR
RANKS0090 RANKS0080
Silver nitrate ≥99.8% LR, Rankem®
Silver nitrate

General lab reagent suitable for routine laboratory usage and classical analysis.


  • Description: Colourless crystals
  • Available in various pack size

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
Merck Index: 12,08661

Order Now

Specification Test Results

Clarity Solution of 5 g in 50 ml ofwater is clear and colourless. Passes test
Guarantee Analysis .
Assay Min. 99.80%
Maximum Limits Of Impurities .
Chloride (Cl) ≤0.001%
Sulphate (So4) ≤0.02%
Substance Not Ppt. By HCl (As Sulphate) Max. 0.0500%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR