Silver nitrate ≥99.8% AR ACS, Rankem®

Supplier: RANKEM
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

S0076 S0077 S0078
RANKS0076EA 12705 INR
RANKS0076 RANKS0077 RANKS0078
Silver nitrate ≥99.8% AR ACS, Rankem®
Silver nitrate

High purity reagents that meet or exceed the ACS reagent Specifications


  • Batch-to-Batch consistency
  • Tested for heavy metals
  • Suitable for advanced chemical testing
  • Available in various pack size


Description: Colourless or white crystals.

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661

Order Now

Specification Test Results

Guarantee Analysis .
Assay (AgNO3) Min. 99.80 %
Identity Passes test
Maximum Limits Of Impurities .
Substance Not Precipitated By HCl Max. 0.01 %
Free Acid Passes test
Chloride (Cl) ≤0.0005%
Copper(Cu) ≤0.0002%
Iron (Fe) ≤0.0002%
Lead ( Pb ) ≤0.001%
Sulphate (SO4) ≤0.002%
Calcium (Ca) Max. 0.001 %
Magnesium (Mg) Max. 0.001 %
Potassium(K) Max. 0.01 %
Sodium (Na) Max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR