Magnesium carbonate heavy 40,0 - 43,5%, powder USP, FCC, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Magnesium carbonate hydroxideMagnesium carbonate basic, heavy

Management of Change (MOC) category = R

MACR5946-32EA 138991.32 INR
MACR5946-32
Magnesium carbonate heavy 40,0 - 43,5%, powder USP, FCC, Macron Fine Chemicals™
Magnesium carbonate heavy
Formula: CH₂Mg₂O₅
MW: 142,63 g/mol
Density: 3,03 g/cm³ (16 °C)
MDL Number: MFCD00011107
CAS Number: 39409-82-0
EINECS: 208-915-9

Order Now

Specification Test Results

GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
Assay (MgO) 40.0 - 43.5 %
Appearance (bulky, white powder) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
Microbiological Passes Test
Soluble Salts ≤ 1.0 %
Acid-Insoluble Substances ≤ 0.05 %
Solubility in Dilute Sulfuric Acid Passes Test
Limit of Calcium (Ca) ≤ 0.45 %
Iron (Fe) ≤ 0.02 %
Calcium Oxide ≤ 0.6 %
Lead (Pb) ≤ 2 mg/kg
Arsenic (As) ≤ 3 ppm
Bulk Density (g/cc) ≥ 0.34
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR