Hematology Analyser BeneSphera™ H31 Plus
Supplier: Avantor
MIND3011D-PA00005EA 324000 INR
MIND3011D-PA00005
Hematology Analyser BeneSphera™ H31 Plus
Clinical Diagnostic Supplies Clinical Analysers
Quantitative hematology analysis and 3 part differential counting for in vitro use in your laboratory. The advanced BeneSphera H31 Plus Hematology Analyzer supports identification of normal patients with system generated parameters and flags patient results that require your attention.
  • CBC + DIFF , 22 paramters, 3 Histograms
  • Throughput upto 40 samples / hour
  • 10.4 inch TFT touch screen
  • Only three reagents needed
  • Detailed flag information
  • Large storage capacity upto 100,000 samples
  • Sample volume is only 9ul which is ideal for pediatric samples
Minimum Size, Maximum Capacity.