Media Bottles, PETG and PET
Supplier: ACCUMAX
AMB-1000-PG AMB-125-PG AMB-2000-PG UAMB-500-PG-S
PFACAMB-1000-PGEA 16185 INR
PFACAMB-1000-PG PFACAMB-125-PG PFACAMB-2000-PG PFACUAMB-500-PG-S
Media Bottles, PETG and PET
Bottles Plastic Bottles
Media bottles for storage of media, biological buffers, and a wide range of reagents. Engineered to perfection, offer a complete package of reliability and convenience.
  • Made of USP Class VI Raw material, with low leachable and extractable
  • Manufactured in 100K clean room and gamma sterile product
  • Non pyrogenic and non cytotoxic
  • Graduated for quick user visibility
  • HDPE closures provided
  • Suitable for frozen storage down upto –70 °C
  • Heat-shrink band around closure and neck for tamper-evident seal
Leak-proof and shatter-proof design, they provide unparalleled protection for your valuable contents. Whether storage or transportation, these bottles excel in every aspect. They are freezer-safe, ensuring optimal preservation of samples even in low-temperature environments. Available in PET and PETG materials and various sizes, offering versatility to suit the specific needs.