Fixed Angle Rotors, iFuge UC02R
Supplier: NEUATION TECHNOLOGIES
UC-149 UC-600 UC-150
NEUAUC-149EA 0 INR
NEUAUC-149 NEUAUC-600 NEUAUC-150
Fixed Angle Rotors, iFuge UC02R
Centrifuges Rotors
Loaded with multiple benefits, iFuge UC02 is manufactured to provide the results under various loads. Having done the best of research, one must not compromise on quality, precision, or design; with a spin of 15000 min⁻¹ and the programmable microprocessor control, UC02 gives superior precision, and detects even a minor imbalance.
  • Auto rotor detection feature
  • Max capacity 4×100 ml (swing out and fixed rotor)
  • Maximum tube capacity 32×6 ml (swing out)
  • Brushless DC motor for maintenance-free long life
  • Single equipment for different work protocols with an appropriate choice of rotors (Swing and Fixed angle)
  • Adjustable ramp and brake speeds from (up to 9 speed selection)
  • User-programmable (user can set up to 99 Programs with 4 line programs)