Rankem HPLC Buffers
Supplier: RANKEM
S0247
P0332
S0241
A0419
S0108
A0341
RANKS0247EA
2001.04
INR
RANKS0247
RANKP0332
RANKS0241
RANKA0419
RANKS0108
RANKA0341
Rankem HPLC Buffers
Buffers
Chromatography Buffers
High purity mobile phase buffers for HPLC improves the reproducibility of the method even at higher concentration
- Significantly improved baseline ensuring reproducible results and higher signal to noise ratio for low concentration samples
- Best performance for gradient analysis
- Least insoluble residues
- Lowest UV absorbance at high concentration
Packaging: Processed with stringent quality checks to ensure accurate analysis and batch to batch reproducibility