Hematology Analyser, BC 780
Supplier: Mindray
MIND3128B-PA00002EA 8580508.47 INR
MIND3128B-PA00002
Hematology Analyser, BC 780
Clinical Diagnostic Supplies Clinical Analysers
BC 780, Advanced Automated Hematology Analyzer
  • BC-700 series integrates an automatic ESR module in a hematology analyzer. It can also generate both CBC & ESR results in one test within 1.5 min.
  • It saves the costs that would otherwise be incurred for the purchase, maintenance, consumables, and storage space of a separate ESR analyzer.
  • Great correlation with traditional Westergren method, this method performs better in quality traceability, repeatability, speed, safety, and level of automation
  • Same QC & Calibrator as in BC-6000 series
  • Combined examination helps to avoid the interferences of dehydration, polycythemia vera and anemia on ESR results