Clinical Chemistry Analyser, BS 240 Pro
Supplier: Mindray
MINDBS-240PROEA 4376059.32 INR
MINDBS-240PRO
Clinical Chemistry Analyser, BS 240 Pro
Clinical Diagnostic Supplies Clinical Analysers
BS 240 Pro, a compact Robust Chemistry Analyzer.
  • Constant throughput of 240 photometric T/H
  • Large and Flexible capacity with upto 100 sample positions and upto 100 reagent positions (50 Fixed + 50 interchangeable)
  • Grating Photometer (100 ul minimum reaction volume)
  • Easy loading and unloading for samples; One key calibrator import