BeneSphera™ TSH MW ELISA - Novo

Supplier: Avantor
BeneSphera™
BENP15001536EA 3519 INR
BENP15001536
BeneSphera™ TSH MW ELISA - Novo
Assays ELISAs
BeneSphera™ TSH Novo Elisa kit is for quantitative determination of TSH (Thyroid Stimulating Hormone) hormone from human serum and plasma.

  • Assay time: 1 hour 30 min
  • Expected range: 0,39 - 6,16 µIU/ml
  • Sensitivity: 0,078 µIU/ml
  • Specificity: No interference found, a total of 170 samples found to have high degree of correlation with TSH calibrators provided
  • Precision: Very low coefficient of variation found in intra-assay and inter-assay reproducibility studies

Enzyme Immunoassay for the quantitative determination of TSH Hormone concentration in human serum/plasma - for invitro diagnostics use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR