BeneSphera™ C Trop I Rapid Card Test

Supplier: Avantor
BeneSphera™
BENP15001327EA 1490.4 INR
BENP15001327
BeneSphera™ C Trop I Rapid Card Test
Assays Immunological Assays
BeneSphera™ C Trop I rapid card test for the detection of cTroponin specific antigen from human serum, plasma, and whole blood.

  • No assay buffer required
  • Can detect cTroponin I Antigen upto 0.5 ng/ml
  • Sensitivity: 99,3%
  • Specificity: 99,5%
  • Test time: 15 to 20 min

BeneSphera™ C Trop I Rapid Card Test is for in vitro diagnostic use only. This test should be used for the detection of Troponin I in whole blood, serum and plasma samples only. Neither the quantitative values nor the rate of increase of cTnI can be determined by this qualitative test.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR