BeneSphera™ LH ELISA Kit

Supplier: Avantor
BeneSphera™
BENP15000977EA 8408.34 INR
BENP15000977
BeneSphera™ LH ELISA Kit
Assays ELISAs
BeneSphera™ LH ELISA Kit for the quantitative determination of LH (Luteinizing hormone) Hormone from human serum and plasma.

  • Quantitative determination by sandwich ELISA
  • Assay time: 1 h 30 min
  • Sensitivity: 0,02 mIU/ml which is equivalent to a sample containing 0,8 mIU/ml LH concentration
  • Specificity: no cross reactivity with interfering substances like follitropin, a- LH sub-unit, TSH and hCG up to 1000 ng/ml

BeneSphera™ LH was studied for intra assay and interassay reproducibility by analysis on pooled control sera. Intra assay reproducibility gave CV of < 5.5% and for interassay was found to be < 6.5%.

Caution: For invitro diagnostics use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR