BeneSphera™ FT4 ELISA Kit

Supplier: Avantor
BeneSphera™
BENP15000970EA 9820.08 INR
BENP15000970
BeneSphera™ FT4 ELISA Kit
Assays ELISAs
BeneSphera™ FT4 ELISA Kit for the quantitative determination of FT4 (Free thyroxine) Hormone from human serum and plasma.

  • Expected range: Adult - 0,8 to 2,0 ng/dl, Pregnancy - 0,76 to 2,24 ng/dl
  • Sensitivity: 0,05 ng/dl
  • Specificity: no interference found, total of 197 samples found to have high degree of correlation with free T4 calibrators provided
  • Precision: very low coefficient of variation found in intra-assay and inter-assay reproducibility studies

Results were compared with a reference radio immunoassay method. Samples were used from euthyroid, hypothyroid and hyperthyroid cases. (Range 0.1 ng/dl - 8.0 ng/dl). The total no. of such specimens was 197.

Caution: For invitro diagnostics use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR