Fittings, Compression Nut Assembly with Ferrule, Jaco Manufacturing®
Supplier: Avantor Fluid Handling
06380-11BLK 06380-41BLK
MFLX06380-11BLKEA 0 INR
MFLX06380-11BLK MFLX06380-41BLK
Fittings, Compression Nut Assembly with Ferrule, Jaco Manufacturing®
Fittings and Connectors
Can handle pressures up to 225 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).