Value Plastics Luer to Hose Barb Fittings
Supplier: Avantor Fluid Handling
45552-04
MFLX45551-01EA 2900 INR
MFLX45551-01 MFLX45552-04 MFLX45555-04
Value Plastics Luer to Hose Barb Fittings
Fittings and Connectors
Premium luer fittings for critical applications.
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options.
All fittings are sold in packs of 100.