Union Fittings, Compression, Elbow
Supplier: Avantor Fluid Handling
31412-07 31412-60
MFLX31412-07EA 2300 INR
MFLX31412-07 MFLX31412-60
Union Fittings, Compression, Elbow
Fittings and Connectors
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).