Adapter Fittings, Female NPT Threaded to Bulkhead, Straight
Supplier: Avantor Fluid Handling
06924-53
MFLX06445-21EA 4700 INR
MFLX06445-21 MFLX06447-41 MFLX06924-53
Adapter Fittings, Female NPT Threaded to Bulkhead, Straight
Fittings and Connectors
Designed for permanent installation.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Bulkhead connectors securely pass through walls and panels, providing a fitting on either side for connections.