Parker Hannifin Threaded Reducing Nipples, NPT(M) x NPT(M)
Supplier: Avantor Fluid Handling
31321-05
MFLX31321-05EA 6000 INR
MFLX31321-05 MFLX31321-06 MFLX31321-07
Parker Hannifin Threaded Reducing Nipples, NPT(M) x NPT(M)
Fittings and Connectors
Create a secure connection with threaded fittings.
  • Constructed of chemically inert PFA
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.