Parker Hannifin Compression Fitting Ferrule Nuts
Supplier: Avantor Fluid Handling
03302-46 03302-51 03302-56
MFLX03302-46EA 1124.36 INR
MFLX03302-46 MFLX03302-51 MFLX03302-56 MFLX03302-44
Parker Hannifin Compression Fitting Ferrule Nuts
Fittings and Connectors
Get a secure seal at higher pressures.
  • Constructed of durable stainless steel
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Compression nuts are required to create a proper seal.