Masterflex® Polypropylene Microcentrifuge Tubes, Avantor®
Supplier: Avantor Fluid Handling
06335-00 06335-01 06335-03 06335-04 06335-05 06335-07 06335-12 06335-15 06335-16 06335-17 06335-18 06335-19 06335-26 06335-31 06335-06
MFLX06335-00EA 0 INR
MFLX06335-00 MFLX06335-01 MFLX06335-03 MFLX06335-04 MFLX06335-05 MFLX06335-07 MFLX06335-12 MFLX06335-15 MFLX06335-16 MFLX06335-17 MFLX06335-18 MFLX06335-19 MFLX06335-26 MFLX06335-31 MFLX06335-06
Masterflex® Polypropylene Microcentrifuge Tubes, Avantor®
Tubes Centrifuge Tubes
Cap locking options for every application.
  • Microcentrifuge tubes with graduations and frosted panels for easy labeling
  • Microbiological cleanroom controlled production (ISO Class 8)
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121 °C for 20 minutes
These Cole-Parmer® polypropylene (PP) microcentrifuge tubes come in three cap lock options for a wide range of applications.
The double-locking caps are ideal when working with valuable samples; they prevent accidental opening during inoculation or storage and are extremely leaktight.
The click-close tubes close firmly and are ideal for heating applications.
The easy-open caps eliminate stress and sore thumbs when opening, and are best for general-purpose use and storage.