Linktech 40AC and 40PP Series Quick-Disconnect Fitting Body, In-Line Hose Barb with Valve
Supplier: Avantor Fluid Handling
41305-09
MFLX41304-08EA 33900 INR
MFLX41304-08 MFLX41305-08 MFLX41305-09
Linktech 40AC and 40PP Series Quick-Disconnect Fitting Body, In-Line Hose Barb with Valve
Fittings and Connectors
Easy to disconnect from 1/4" flow line with one hand.
  • Withstands pressures from vacuum to 120 psi (8.3 bar)
  • Interchangeable with other similar industry thumb-latch fittings
Just press the metal thumb-latch to connect and disconnect fitting halves. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized.
Mix and match valved and straight-through (nonvalved) fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.