L-K Industries Transportable Centrifuge
Supplier: Avantor Fluid Handling
17700-51 17700-42 17700-43 17700-44 17700-45 17700-46 17700-47 17700-49 17700-50
MFLX17700-51EA 0 INR
MFLX17700-51 MFLX17700-42 MFLX17700-43 MFLX17700-44 MFLX17700-45 MFLX17700-46 MFLX17700-47 MFLX17700-49 MFLX17700-50
L-K Industries Transportable Centrifuge
Centrifuges Benchtop Centrifuges
Get fast test results on crude oil haulers.
  • Rugged design offers longer life and less downtime
  • Compact design makes installation and maintenance easy
These transportable centrifuges are ideal for determining sediment and water content on crude oil haulers. The centrifuges are available in analog and digital models with a heated or nonheated configuration, and either a 2- or 4-place Trunnion arm.
Each model features a sturdy outer steel case with reinforced mounting rails for outdoor use and variable-speed motor controller with epoxy-coated electronics.
More Product Information