Pentair Quick-Change Filtration System Accessories
Supplier: Avantor Fluid Handling
01507-42
MFLX01507-42EA 6788.81 INR
MFLX01507-42
Pentair Quick-Change Filtration System Accessories
Filters Cartridge Filter Systems
Optimize your Pentair Quick-Change Filtration Systems.
  • Choose from replacement cartridges and bracketed heads for your your Pentair Quick-Change Filtration Systems
The quick-change filtration systems offer a complete solution in one simple design—several cartridges will meet your water quality needs. Easily change the cartridge filter with a quarter-turn of the filter. The filter head features a built-in auto shut-off when cartridge is removed, eliminating the need for additional shutoff valves in your filter system. Connections on inlets and outlets are available with 1/4" NPT fittings, or 1/4" and 3/8" John Guest® push-to-connect fittings.
Each quick-change system includes a cartridge, filter head, and mounting bracket for use as a single unit; single cartridge systems are NSF certified. You can also combine cartridges in a series of up to three filters with the optional double and triple bracketed heads (sold separately).