PharmaFluor FEP Tubing
Supplier: Avantor Fluid Handling
16356-48
MFLX15943-81EA 60700 INR
MFLX15943-81 MFLX15943-83 MFLX15943-84 MFLX15943-85 MFLX15943-86 MFLX15943-87 MFLX15943-88 MFLX16356-48
PharmaFluor FEP Tubing
Tubing Transfer Tubing
Applications: Sterile filling and dispensing, diagnostic equipment, general laboratory use and Analytical & process equipments in biotechnology and biopharmaceutical.
Characteristics: Chemically inert, ultra pure, non-cytotoxic, non-pyrogenic and non-hemolytic, extreamly low sorption, fully autoclavable and sterilizable
Certification: Complies USP Class VI & ISO Standards and has a Master File with U.S. Food and Drug Administration
Temperature range: -73 °C to 204 °C(-100° F to 400° F)
Sterilise: Autoclave, ethylene oxide or gamma irradiation
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.