Masterflex® Ismatec® Wash-Down Process Pumps, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MFLX78003-10EA 0 INR
MFLX78003-10
Masterflex® Ismatec® Wash-Down Process Pumps, Avantor®
Pumps Peristaltic Pumps
Perfect for lab and process automation.
  • Flow rate 0.45 to 3400 ml/min - flow rate depends on drive rpm, pump head, and tubing size
  • IP65 rated housing
  • Offers remote-control capabilities
These pumps are ideal for dosing, filling, and batch programming. The aluminum pump head with stainless steel rotors disassembles readily for cleaning. Speed setting is rpm. Flow rate setting is µL/min or ml/min.
Washdown Process Pumps are easily programmable via the seven-button keypad. View rpm and direction on the easy-to-read LED. Remotely control start/stop functions via a 5 VDC, 10 VDC, 0 to 20 mA or 4 to 20 mA input.
Delivery information: Includes drive and a 6-ft. (1.8-m) cord with plug.