C-Flex ULTRA Tubing
Supplier: Avantor Fluid Handling
15943-29
MFLX15943-25EA 11600 INR
MFLX15943-25 MFLX15943-28 MFLX15943-33 MFLX15943-34 MFLX15943-29
C-Flex ULTRA Tubing
Tubing Transfer Tubing
Applications: Pharmaceutical and biotechnology applications. Cell culture media and fermentation, and single-use assemblies.
Characteristics: Exceptional pump life, low spallation and permeability, smoother outer surface, excellent clarity, thermally sealable and weldable. Non-pyrogenic and non-cytotoxic. Animal-derived component-free. Contains no DEHP or other phthalate plasticizers.
Certifications: Resin is FDA compliant; meets USP Class VI and EP requirements. Tubing is REACH-compliant. Tubing is manufactured according to GMP.
Temperature range: –50 to 110 °C (–58 to 230 °C)
Sterilize: Autoclave or gamma irradiation.