Masterflex® Value Plastics Luer Fittings and Adapters 61470, Avantor®
Supplier: Avantor Fluid Handling
45551-00 45552-38 45552-01 45552-42 45552-00 45555-31
MFLX45551-00EA 3300 INR
MFLX45551-00 MFLX45552-38 MFLX45509-01 MFLX45552-02 MFLX45555-74 MFLX45552-01 MFLX45555-02 MFLX45552-42 MFLX45509-03 MFLX45552-00 MFLX45555-31 MFLX45552-60 MFLX45555-56
Masterflex® Value Plastics Luer Fittings and Adapters 61470, Avantor®
Fittings and Connectors
Premium luer fittings for critical applications.
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety ofMore Product Information