Hayward Diaphragm Valves with True Union Connections, PVC
Supplier: Avantor Fluid Handling
07390-55
MFLX07390-55EA 80500 INR
MFLX07390-55
Hayward Diaphragm Valves with True Union Connections, PVC
Valves Diaphragm Valves
Designed for use in corrosive environments.
  • Multi-turn handle gives you precise control
  • Position indicator shows exact location of valve
  • Self draining allows for little dead volume in valve
  • PVC body and EPDM diaphragm
These Hayward diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 140 °F (60 °C) and feature true-union connections.