Two Way Solenoid Valves for Industrial Applications
Supplier: Avantor Fluid Handling
08610-04 08610-06
MFLX08610-04EA 45397.82 INR
MFLX08610-04 MFLX08610-06
Two Way Solenoid Valves for Industrial Applications
Valves Solenoid Valves
Don’t worry about leaks.
  • Direct acting, normally closed (NC)
  • Encapsulated plug-in coils
  • Buna N seat material
  • DIN 43-650 electrical connection
  • IP65 and NEMA 4 rated connection
Forged brass-body solenoid valve is designed for use in low- and medium-vacuum systems, fuel oil and gas burners, oxygen and acetylene equipment, and water, air, and light oil systems.