Pure-Fit SPT-60 Sanitary Silicone Tubing
Supplier: Avantor Fluid Handling
94703-33 94703-26
MFLX16357-13EA 51000 INR
MFLX16357-13 MFLX94703-30 MFLX94703-31 MFLX94703-32 MFLX94703-34 MFLX94703-36 MFLX94703-38 MFLX94703-44 MFLX94703-49 MFLX94703-33 MFLX94703-26 MFLX16356-99 MFLX16357-40
Pure-Fit SPT-60 Sanitary Silicone Tubing
Tubing Transfer Tubing
Pure-Fit SPT-60 tubing features ultra-smooth inner surfaces to inhibit protein binding as well as bacterial growth. The excellent quality of this tubing ensures easy sterilization, too. The platinum-cured silicone elastomer material used in the tubing is pharmaceutical grade with minimal extractables.
  • Usable from −62 to 260 °C
  • May be sterilized by autoclaving, ethylene oxide, or gamma irradiation
Tubing material meets USP Class VI and EP 3.1.9 requirements and is nontoxic and nonpyrogenic. Manufactured in a Class 10,000 clean room, all tubing offers complete lot traceability.