Masterflex® Union Fittings, Compression, Straight, Avantor®
Supplier: Avantor Fluid Handling
06381-50 06382-15 30802-95 30803-04 30803-05 30803-06 30805-04 30805-05
MFLX06381-50EA 11800 INR
MFLX06381-50 MFLX06382-15 MFLX30802-95 MFLX30803-04 MFLX30803-05 MFLX30803-06 MFLX30805-04 MFLX30805-05
Masterflex® Union Fittings, Compression, Straight, Avantor®
Fittings and Connectors
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).