UL-Listed Air-Operated Double Diaphragm Pump
Supplier: Warren Rupp
MFLX74440-90EA 284478.63 INR
MFLX74440-90
UL-Listed Air-Operated Double Diaphragm Pump
Pumps Air-Operated Pumps
Designed to meet UL79 Standards for handling flammable liquids.
  • Fully groundable to prevent static discharge
  • Handles abrasive and shear-sensitive materials
  • Self-priming up to 24 ft of water
  • Deadhead against closed discharge without damage to the pump
These UL-Listed Air-Operated Double Diaphragm Pumps are constructed of all aluminum with approved Buna-N or virgin PTFE diaphragm, making them ideal for pumping flammable liquids.