PFA Compression Male Threaded Adapter Elbows
Supplier: Avantor Fluid Handling
31320-14 31320-15 31320-16 31320-19 31320-77 31321-70 31321-71
MFLX31320-17EA 8738.3 INR
MFLX31320-17 MFLX31320-20 MFLX31320-75 MFLX31320-78 MFLX31320-80 MFLX31320-14 MFLX31320-15 MFLX31320-16 MFLX31320-19 MFLX31320-77 MFLX31321-70 MFLX31321-71 MFLX31320-18 MFLX31320-76 MFLX31320-79 MFLX31320-81
PFA Compression Male Threaded Adapter Elbows
Fittings and Connectors
Part of our vast assortment of compression fittings.
  • Compression x NPT(M)
  • Sterilized via autoclave or ethylene oxide (ETO)
  • Constructed of durable PFA
Count on this this quality-made compression threaded adapter elbow to suit your needs.