Spirax Sarco Cast Iron Air and Gas Eliminators
Supplier: Avantor Fluid Handling
07600-92 07600-94
MFLX07600-92EA 85300 INR
MFLX07600-92 MFLX07600-94
Spirax Sarco Cast Iron Air and Gas Eliminators
Filters Gas Filters
Improve the circulation of pressurized liquids by eliminating air and other non-condensable gases.
  • Mount at high points to provide immediate removal of air and gases
  • EPDM seals ensure positive shut-off
  • Cast-iron body with stainless steel internal components — can be serviced without disturbing the inlet piping connection
Typical applications:
Use for both hot and cold liquid services
Cold water lines
Suction lines to pumps
Mixing tanks
Condensate return lines
Cool water lines on air compressors
Water storage tanks