Masterflex® Snap Cap Microcentrifuge tubes, Avantor®
Supplier: Avantor Fluid Handling
06333-51 06333-67 06333-68
MFLX06333-51EA 0 INR
MFLX06333-51 MFLX06333-67 MFLX06333-68
Masterflex® Snap Cap Microcentrifuge tubes, Avantor®
Tubes Centrifuge Tubes
General purpose microcentrifuge tubes with SnapLock cap.
Cole-Parmer microcentrifuge tubes include an attached cap. All tubes feature a frosted writing surface on the tube and are graduated. Tubes are available in assorted colors (tube and cap) for quick sample identification. Boil-proof tubes are guaranteed to stay closed, even during boiling or autoclaving. All tubes are certified RNase-, DNase- and endotoxin-free for highest sample integrity