Brass Female/Male Mini Manual Ball Valves
Supplier: Avantor Fluid Handling
30533-22 30533-20 30533-24
MFLX30533-22EA 4217.94 INR
MFLX30533-22 MFLX30533-20 MFLX30533-24
Brass Female/Male Mini Manual Ball Valves
Valves Ball Valves
Don’t worry about leaks.
  • Full flow design ensures maximum operating efficiency
  • Blow-out proof stems
Mini brass ball valves are designed for use in confined and hard-to-reach areas and are ideal for use on water and air service lines.