Automatic-Demand Diaphragm Pumps
Supplier: Avantor Fluid Handling
75420-10 75420-20 75420-37 75420-17
MFLX75420-10EA 62900 INR
MFLX75420-10 MFLX75420-20 MFLX75420-37 MFLX75420-17
Automatic-Demand Diaphragm Pumps
Pumps Diaphragm Pumps
Built-in pressure demand switch shuts pump off at maximum pressure.
  • Three-chamber design provides smooth, pulse-free flow
  • Flexible diaphragm design allows pump to run dry without damage.
  • Compact — mount in any position
Model 75420-20 has an 80 psi internal bypass.
What's included: 115 VAC models include a 6-ft power cord; 12 VDC models have 6" leads.