Hayward Electrically Actuated True Union Ball Valves
Supplier: Avantor Fluid Handling
01347-00 01347-10 01347-22 01347-04 01347-20 01347-30
MFLX01347-00EA 122000 INR
MFLX01347-00 MFLX01347-10 MFLX01347-22 MFLX01347-04 MFLX01347-20 MFLX01347-30
Hayward Electrically Actuated True Union Ball Valves
Valves Ball Valves
Ideal for remote control of process fluids.
  • Precision machined stems specifically designed for actuated valves
  • Adjustable and reversible PTFE(R) PTFE seats
  • One-piece PVC or CPVC body construction
  • Includes two Viton(R) seals for double leakproof protection
  • Remote control capabilities via an auxiliary limit switch
  • Thermal overload circuit protects the motor from burnout and overheating
  • Easy connections via a wiring terminal strip
  • Unique mounting feature allows for easy conversion to manual operation
  • Mechanical brake for precise cycle control
  • Actuator comes mounted on the valve
  • NEMA 4X polypropylene housing stands up to harsh operating conditions
Choose between PVC and CPVC valve materials for a wide range of applications.