Two-Way PVC Ball Valves, Female NPT
Supplier: Avantor Fluid Handling
07387-22 07387-62 07387-42
MFLX07387-22EA 3846.42 INR
MFLX07387-22 MFLX07387-62 MFLX07387-42
Two-Way PVC Ball Valves, Female NPT
Valves Ball Valves
Create a secure pipe threaded connection.
  • Constructed of durable PVC
Control flow with the 90° rotating handle. Valves have seals made of Viton® fluoroelastomer and seats made of PTFE. Maximum pressure is 150 psi at 32 °F (10.3 bar at 0 °C); maximum temperature is 140 °F (60 °C at 1.7 bar).