Masterflex® Ismatec® Analog Variable-Speed Peristaltic Pump Drives, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MFLX78002-12EA 474000 INR
MFLX78002-12
Masterflex® Ismatec® Analog Variable-Speed Peristaltic Pump Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
No more difficult pump head mounting.
  • Mount pump heads quickly with turn-and-lock mounting system
Quick-couple drives feature a turn-and-lock mounting system that makes installing pump heads fast and easy. Select from the interchangeable pump heads on page 1607 of our 2013/2014 virtual catalog to achieve flow rates from 0.003 to 3600 ml/min. Control speed remotely on all models via analog signal (0 to 5 VDC, 0 to 10 VDC, 0 to 20 mA, or 4 to 20 mA). Start/stop pump or change flow direction remotely via DB15 female contact enclosure.
Adjust speed in 0.1% steps with the potentiometer. Use MAX/SLOW toggle switch to prime and flush or to decrease speed to prevent overflow.
See link below for tubing options or for personal assistance call our Application Specialists.
Delivery information: Includes 6-ft (1.8-m) cord with plug.