Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
MFLX07516-00EA 0 INR
MFLX07516-00 MFLX07516-40 MFLX07518-60
Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Save time with fast tubing changes and reduced maintenance.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Select either stainless steel (SS) or cold-rolled steel (CRS) rotor—choose CRS for economy or SS for greater durability and chemical resistance
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
This Easy-Load® pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18.
Delivery information: Includes single-channel mounting hardware and a 15" (38-cm) length of silicone pump tubing.