Conical-Bottom Reusable Plastic Tubes
Supplier: Avantor Fluid Handling
06325-10 06326-10 06326-50 06325-50
MFLX06325-10EA 0 INR
MFLX06325-10 MFLX06326-10 MFLX06326-50 MFLX06325-50
Conical-Bottom Reusable Plastic Tubes
Tubes Centrifuge Tubes
Molded-in graduation simplify measurements.
  • Designed for use up to 6000 × g in refrigerated or non-refrigerated centrifuges
  • Autoclavable
The conical-bottle centrifuge tubes feature molded-in graduations allowing you to easily read measurements. They are designed for low-speed centrifugation up to 6000 ×g in refrigerated or non-refrigerated centrifuges. Please note, tubes must be filled to at least 80% capacity for proper function. The conical bottoms concentrate pellet in a small area for easy isolation and retrieval. Available in either polypropylene copolymer (PPCO) or polycarbonate (PC).
The translucent PPCO tubes are compatible with a wide range of lab reagents and offer excellent chemical resistance. The PC tubes feature outstanding mechanical strength and clarity.