VWR®, Volumetric Flask, USP, with PP Cap
Supplier: VWR Collection
612-6842EA 2347.63 INR
612-6842 612-6843 612-6844 612-6845 612-6846 612-6847 612-6848 612-6849 612-6850 612-6851
VWR®, Volumetric Flask, USP, with PP Cap
Flasks Volumetric Flasks
Manufactured from borosilicate 3.3 glass, clear with PP stopper, offering excellent thermal and chemical resisteance. VWR® Volumetric flasks are calibrated to to measure the amount of solvent contained.
  • Calibrated at 20° C
  • Complies with ASTM E288-03 Class A
  • Manufactured from 3.3 borosilicate glass for corrosion-free performance
  • Allows accurate volume measurment and determination
  • Blue color inscriptions
  • Individual certification
Applications include accurate measurement of liquid amounts, storage, and preparation of standard solutions.