VWR®, Examination Gloves, Latex, Powder-Free
Supplier: VWR Collection
VWRA112-5439EA 5966.8 INR
VWRA112-5439 VWRA112-5440 VWRA112-5432 VWRA112-5433 VWRA112-5438 VWRA112-5441 VWRA112-5442 VWRA112-5443 VWRA112-5434 VWRA112-5435 VWRA112-5436 VWRA112-5437 VWRA112-5444 VWRA112-5445 VWRA112-5446 VWRA112-5447 VWRA112-5448 VWRA112-5449
VWR®, Examination Gloves, Latex, Powder-Free
Gloves
VWR® Latex powder-free gloves are designed for use in controlled enviorement applications.
  • Natural rubber latex for superior tactile sensitivity and comfort
  • Fully textured surface for better grip
  • Powder-free gloves reduce chances of cross contamination
  • Ambidextrous ergonomic design for convenient donning
Recommended for sample collection and processing, testing, maintenance in life science and chemical industries. Gamma sterilised option is also available.
QR coded pouch packaging with easy access of technical data sheet and batch specific COA.
Certifications: Gloves comply to ASTM D3578 and EN 455 standards.
Packaging: Gloves are packaged 50 pair per pack.
Caution: This product contains natural rubber latex, which may cause an allergic reaction.