EDTA disodium salt dihydrate, Molecular Biology Grade

Supplier: VWR Collection

Synonyms: Ethylenediaminetetraacetic acid disodium dihydrateEthylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateEthylenediaminetetraacetic acid, disodium salt, dihydrateEthylenediaminetetraacetic acid disodium salt dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIKestranal® 2S dihydrateEdetic acid disodium salt dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrateSequestrene Na₂

VWR®
VWRA06190112EA 9600 INR
VWRA06190112
EDTA disodium salt dihydrate, Molecular Biology Grade
EDTA disodium salt dihydrate

VWR Delivered High quality Reagents & biochemicals, combined with innovative products that gives optimum results , save time & simplify protocols


  • VWR Lifescience Molecular Biology grade Reagents & Biochemicals.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372,24 g/mol
MDL Number: MFCD00150037
CAS Number: 6381-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR