VWR®, Snap Ring Vials, ND11, with Closures
Supplier: VWR Collection
548-2576EA 1512.5 INR
548-2576 548-2577 548-2574 548-2575
VWR®, Snap Ring Vials, ND11, with Closures
Vials
These vials are for use on instruments from the following manufacturers: Agilent, CTC, DANI, Dionex, Jasco, Shimadzu, Spark, Thermo Scientific, Varian, VWR®/Hitachi, Waters®, more.
  • Type-1 borosilicate glass
  • Universally usable vials for almost all autosamplers, even for those with robotic handling
  • Vials can also be crimped with a standard 11 mm aluminium crimp seal, as the two snap-ring lips have the same height as a crimp neck
  • Wide opening enables easy filling with viscous materials
This vial/closure system is recommended for HPLC applications only.
Certifications: Each pack contains a Certificate of Confirmity (COC).
Delivery information: The combopacks of vials and caps contain vials and septa caps - 100 pieces each.
Packaging: Vials are supplied completely shrink-wrapped to ensure origin, purity and transport safety.